Learn More
Medchemexpress LLC HY-133024 5mg Medchemexpress, Flumexadol CAS:30914-89-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1330245MG
Encompass_Preferred
Medchemexpress, HY-133024 5mg Flumexadol CAS:30914-89-7 Flumexadol is an orally active non-narcotic analgesic. Flumexadol is a selective and affinity 5-HT2C receptor agonist with a Ki of 25 nM for the (+)-enantiomer of Flumexadol, and is 40-fold selective over the 5-HT2A receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9845