Learn More
AdipoGen Flumiclorac
SDP
Supplier:  AdipoGen CDXF0085M250
Encompass_Preferred
Chemical. CAS 87547-04-4. Formula C16H13ClFNO5. MW 353.73. Synthetic Herbicide. Mode of action is similar to herbicides that inhibit the protoporphyrinogen oxidase Protox enzyme in susceptible plant species that leads to an uncontrolled nonenzymatic oxidation of protoporphyrinogen IX Protogen to protoporphyrin IX Proto and successive necrosis.
Catalog No. 50-256-0883