Learn More
Enzo Life Sciences Flunarizine . dihydrochloride (1g). CAS: 30484-77-6
SDP
Supplier:  Enzo Life Sciences ALX550268G001
Encompass_Preferred
Selectively blocks the low threshold (T-type) calcium channel in rat hippocampal CA1 pyramidal neurons. Inhibits Ca2+-influx-induced vasoconstriction of peropheral and cerebral blood vessels with little effect on myogenic activity and Ca2+-depenent myocardial contractility. Alternative name: 1-[bis(4-Fluorophenyl)methyl]-4-(3-phenyl-2-propenyl)piperazine . 2HCl. Purity: ≥99% (HPLC). Appearance: White solid. Solubility: Soluble in DMSO (25mg/ml), 100% ehanol (5mg/ml) or chloroform:methanol (1:1; 50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9517