Learn More
Medchemexpress LLC Flunarizine dihydrochloride | 30484-77-6 | MFCD00058198 | 99.9% | 500 MG
SDP
Catalog No. 501639359
Encompass_Preferred
Flunarizine dihydrochloride is a potent dual Na+/Ca2+ channel (T-type) blocker. It also acts as a D2 dopamine receptor antagonist, exhibiting anticonvulsive and antimigraine activities, along with peripheral vasodilator effects.
  • Potent dual Na+/Ca2+ channel (T-type) blocker
  • D2 dopamine receptor antagonist
  • Demonstrates anticonvulsive activity
  • Exhibits antimigraine activity
  • Provides peripheral vasodilator effects
  • Blocks sodium currents and calcium currents (INa and ICa)
  • Protects against LPS-induced acute lung injury
  • Suppresses LPS-induced cell influx, protein leakage, and inflammatory cytokines release
  • Inhibits pulmonary inflammation
Catalog No. 50-163-9359 Supplier Medchemexpress LLC Supplier No. HYB0358A500MG
Flunarizine dihydrochloride is a potent dual Na+/Ca2+ channel (T-type) blocker. It also acts as a D2 dopamine receptor antagonist, exhibiting anticonvulsive and antimigraine activities, along with peripheral vasodilator effects.
  • Potent dual Na+/Ca2+ channel (T-type) blocker
  • D2 dopamine receptor antagonist
  • Demonstrates anticonvulsive activity
  • Exhibits antimigraine activity
  • Provides peripheral vasodilator effects
  • Blocks sodium currents and calcium currents (INa and ICa)
  • Protects against LPS-induced acute lung injury
  • Suppresses LPS-induced cell influx, protein leakage, and inflammatory cytokines release
  • Inhibits pulmonary inflammation