Learn More
Medchemexpress LLC Flunarizine dihydrochloride | 30484-77-6 | 99.9% | 1 G
SDP
Catalog No. 5000407116
Encompass_Preferred
Flunarizine dihydrochloride is a potent dual Na+/Ca2+ channel (T-type) blocker and a D2 dopamine receptor antagonist. It exhibits anticonvulsive, antimigraine, and peripheral vasodilator effects.
  • Potent dual Na+/Ca2+ channel (T-type) blocker
  • D2 dopamine receptor antagonist
  • Exhibits anticonvulsive activity
  • Exhibits antimigraine activity
  • Exhibits peripheral vasodilator effects
Catalog No. 50-004-07116 Supplier Medchemexpress LLC Supplier No. HYB0358A1G
Flunarizine dihydrochloride is a potent dual Na+/Ca2+ channel (T-type) blocker and a D2 dopamine receptor antagonist. It exhibits anticonvulsive, antimigraine, and peripheral vasodilator effects.
  • Potent dual Na+/Ca2+ channel (T-type) blocker
  • D2 dopamine receptor antagonist
  • Exhibits anticonvulsive activity
  • Exhibits antimigraine activity
  • Exhibits peripheral vasodilator effects