Learn More
Medchemexpress LLC Flunarizine (dihydrochloride) | 30484-77-6 | 99.9% | 1 ML
SDP
Catalog No. 5000407172
Encompass_Preferred
Flunarizine dihydrochloride is a potent dual Na+/Ca2+ channel (T-type) blocker and a D2 dopamine receptor antagonist. It exhibits anticonvulsive and antimigraine activity, as well as peripheral vasodilator effects. This product is for research use only, not for sale to patients.
  • Potent dual Na+/Ca2+ channel (T-type) blocker
  • D2 dopamine receptor antagonist
  • Exhibits anticonvulsive and antimigraine activity
  • Exhibits peripheral vasodilator effects
  • Harmful if swallowed
  • Store at 4°C, sealed, away from moisture
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture
Catalog No. 50-004-07172 Supplier Medchemexpress LLC Supplier No. HYB0358A10MM/1ML
Flunarizine dihydrochloride is a potent dual Na+/Ca2+ channel (T-type) blocker and a D2 dopamine receptor antagonist. It exhibits anticonvulsive and antimigraine activity, as well as peripheral vasodilator effects. This product is for research use only, not for sale to patients.
  • Potent dual Na+/Ca2+ channel (T-type) blocker
  • D2 dopamine receptor antagonist
  • Exhibits anticonvulsive and antimigraine activity
  • Exhibits peripheral vasodilator effects
  • Harmful if swallowed
  • Store at 4°C, sealed, away from moisture
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture