Learn More
Medchemexpress LLC Flunarizine dihydrochloride | 30484-77-6 | 99.9% | 10 G
SDP
Catalog No. 5000407277
Encompass_Preferred
Flunarizine dihydrochloride is a potent dual Na+/Ca2+ channel (T-type) blocker and a D2 dopamine receptor antagonist. It demonstrates anticonvulsive and antimigraine activity, as well as peripheral vasodilator effects.
  • Potent dual Na+/Ca2+ channel (T-type) blocker
  • D2 dopamine receptor antagonist
  • Shows anticonvulsive and antimigraine activity
  • Provides peripheral vasodilator effects
  • For research use only
Catalog No. 50-004-07277 Supplier Medchemexpress LLC Supplier No. HYB0358A10G
Flunarizine dihydrochloride is a potent dual Na+/Ca2+ channel (T-type) blocker and a D2 dopamine receptor antagonist. It demonstrates anticonvulsive and antimigraine activity, as well as peripheral vasodilator effects.
  • Potent dual Na+/Ca2+ channel (T-type) blocker
  • D2 dopamine receptor antagonist
  • Shows anticonvulsive and antimigraine activity
  • Provides peripheral vasodilator effects
  • For research use only