Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flunarizine (dihydrochloride) | 30484-77-6 | 99.9% | 5 G
SDP

Catalog No. 5000407173
Encompass_Preferred

Flunarizine dihydrochloride is a potent dual Na+/Ca2+ channel (T-type) blocker and a D2 dopamine receptor antagonist. It exhibits anticonvulsive and antimigraine activity, alongside peripheral vasodilator effects.

  • Potent dual Na+/Ca2+ channel (T-type) blocker
  • D2 dopamine receptor antagonist
  • Exhibits anticonvulsive and antimigraine activity
  • Shows peripheral vasodilator effects
  • Target: T-type calcium channel and D2 receptor
  • Solubility in DMSO: 50 mg/mL (104.73 mM)
  • Solubility in H2O: 1 mg/mL (2.09 mM)

Catalog No. 50-004-07173 Supplier Medchemexpress LLC Supplier No. HYB0358A5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flunarizine dihydrochloride is a potent dual Na+/Ca2+ channel (T-type) blocker and a D2 dopamine receptor antagonist. It exhibits anticonvulsive and antimigraine activity, alongside peripheral vasodilator effects.

  • Potent dual Na+/Ca2+ channel (T-type) blocker
  • D2 dopamine receptor antagonist
  • Exhibits anticonvulsive and antimigraine activity
  • Shows peripheral vasodilator effects
  • Target: T-type calcium channel and D2 receptor
  • Solubility in DMSO: 50 mg/mL (104.73 mM)
  • Solubility in H2O: 1 mg/mL (2.09 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.