Learn More
Medchemexpress LLC Flunarizine (Z)-isomer | 693765-11-6 | MFCD32182831 | 25mg
SDP
Catalog No. 5000269242
Encompass_Preferred
(Z)-Flunarizine is the (Z)-isomer of Flunarizine (HY-B0358) Flunarizine is a potent dual Na /Ca2 channel (T-type) blocker Flunarizine is a D2 dopamine receptor antagonist Flunarizine shows anticonvulsive and antimigraine activity and peripheral vasodilator effects[1]
Catalog No. 50-002-69242 Supplier Medchemexpress LLC Supplier No. HYZ817625MG
(Z)-Flunarizine is the (Z)-isomer of Flunarizine (HY-B0358) Flunarizine is a potent dual Na /Ca2 channel (T-type) blocker Flunarizine is a D2 dopamine receptor antagonist Flunarizine shows anticonvulsive and antimigraine activity and peripheral vasodilator effects[1]