Learn More
Cayman Chemical Flunarizine (hydrochloride), Purity: ≥98%, CAS: 30484-77-6, 1 g
SDP
Catalog No. 502983257
Encompass_Preferred
A calcium antagonist that acts as a potent dilator of peripheral vessels; reduces calcium fluxes in vascular smooth muscle by blocking both voltage-dependent calcium channels and receptor-operated channels
Catalog No. 50-298-3257 Supplier Cayman Chemical Supplier No. 203851 G
A calcium antagonist that acts as a potent dilator of peripheral vessels; reduces calcium fluxes in vascular smooth muscle by blocking both voltage-dependent calcium channels and receptor-operated channels