Learn More
Cayman Chemical Flunisolide, Purity: ≥95%, CAS: 3385-03-3, 50 mg
SDP
Catalog No. 502994838
Encompass_Preferred
A fluorinated corticosteroid receptor agonist; induces reversible pyloric hyalinization in CD-1 mice following oral administration at a dose of 200 μg/kg per day
Catalog No. 50-299-4838 Supplier Cayman Chemical Supplier No. 2384250 MG
A fluorinated corticosteroid receptor agonist; induces reversible pyloric hyalinization in CD-1 mice following oral administration at a dose of 200 μg/kg per day