Learn More
Cayman Chemical FlunIxIn-d3 10mg
SDP
Catalog No. 502911006
Encompass_Preferred
An internal standard for the quantification of flunixin by GC- or LC-MS
Catalog No. 50-291-1006 Supplier Cayman Chemical Supplier No. 3505610 MG
An internal standard for the quantification of flunixin by GC- or LC-MS