Learn More
Med Vet International Flunixin Meglumine 50mg/mL, 250mL
Catalog No. 502827949
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Horse: Flunixin Injection is recommended for the alleviation of inflammation and pain associated with musculoskeletal disorders in the horse. It is also recommended for the alleviation of visceral pain associated with colic in the horse. Cattle: Flunixin Injection is indicated for the control of pyrexia associated with bovine respiratory disease, endotoxemia and acute bovine mastitis. Flunixin Injection is also indicated for the control of inflammation in endotoxemia.
Catalog No. 50-282-7949 Supplier Med Vet International Supplier No. RXFLUNIX250
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Horse: Flunixin Injection is recommended for the alleviation of inflammation and pain associated with musculoskeletal disorders in the horse. It is also recommended for the alleviation of visceral pain associated with colic in the horse. Cattle: Flunixin Injection is indicated for the control of pyrexia associated with bovine respiratory disease, endotoxemia and acute bovine mastitis. Flunixin Injection is also indicated for the control of inflammation in endotoxemia.