Learn More
Apexbio Technology LLC Flunixin Meglumin 42461-84-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062355
Encompass_Preferred
Flunixin meglumine (CAS 42461-84-7) is a potent non-steroidal anti-inflammatory drug (NSAID) known to inhibit cyclooxygenase (COX) enzymes It exerts its biological effects primarily by blocking prostaglandin synthesis through non-selective inhibition of COX isoforms exhibiting an IC50 of approximately 1 nM Preclinical studies indicate analgesic and antipyretic properties in rodent and primate pain models Flunixin meglumine is utilized experimentally to modulate inflammatory responses particularly in veterinary models involving ischemic enteritis endotoxemia and colic
Catalog No. 50-000-62355 Supplier Apexbio Technology LLC Supplier No. B14455.1
 Restricted Item
Add to Cart
Add to Cart
Flunixin meglumine (CAS 42461-84-7) is a potent non-steroidal anti-inflammatory drug (NSAID) known to inhibit cyclooxygenase (COX) enzymes It exerts its biological effects primarily by blocking prostaglandin synthesis through non-selective inhibition of COX isoforms exhibiting an IC50 of approximately 1 nM Preclinical studies indicate analgesic and antipyretic properties in rodent and primate pain models Flunixin meglumine is utilized experimentally to modulate inflammatory responses particularly in veterinary models involving ischemic enteritis endotoxemia and colic