Learn More
Medchemexpress LLC Flunixin (meglumine) | 42461-84-7 | 99.65% | 1 ML
SDP
Catalog No. 5000407158
Encompass_Preferred
Flunixin meglumine is a cyclooxygenase (COX) inhibitor exhibiting anti-inflammatory effects. It inhibits lipopolysaccharide-induced inducible nitric oxide synthase (iNOS) activity and nuclear factor kappa B (NfκB) activation in RAW 264.7 murine macrophages. In vivo, it inhibits carrageenan-induced exudate PGE2 and serum TXB2 generation.
Catalog No. 50-004-07158 Supplier Medchemexpress LLC Supplier No. HYB038610MM/1ML
Flunixin meglumine is a cyclooxygenase (COX) inhibitor exhibiting anti-inflammatory effects. It inhibits lipopolysaccharide-induced inducible nitric oxide synthase (iNOS) activity and nuclear factor kappa B (NfκB) activation in RAW 264.7 murine macrophages. In vivo, it inhibits carrageenan-induced exudate PGE2 and serum TXB2 generation.