Learn More
Cayman Chemical Flunixin (meglumine), Purity: ≥98%, CAS: 42461-84-7, 100 mg
SDP
Catalog No. 502933891
Encompass_Preferred
A COX inhibitor (IC50s = 3.24 and 0.55 µM for COX-1 and COX-2, respectively); inhibits carrageenan-induced PGE2 and TXB2 production in sheep exudate and serum, respectively, at 1.1 mg/kg; has anti-inflammatory, analgesic, and antipyretic activity in horses at 1.1 mg/I/kg
Catalog No. 50-293-3891 Supplier Cayman Chemical Supplier No. 26644100 MG
A COX inhibitor (IC50s = 3.24 and 0.55 µM for COX-1 and COX-2, respectively); inhibits carrageenan-induced PGE2 and TXB2 production in sheep exudate and serum, respectively, at 1.1 mg/kg; has anti-inflammatory, analgesic, and antipyretic activity in horses at 1.1 mg/I/kg