Learn More
Apexbio Technology LLC Fluo-3-AM-Calcium-Assay-Kit
SDP
Catalog No. 5000567948
Encompass_Preferred
Fluo-3 AM Calcium Assay Kit is a widely used tool in biomedical research for monitoring calcium ion concentrations in various cellular processes Originating from the category of fluorescent calcium indicators this kit operates by permeating cell membranes and then being hydrolyzed by intracellular esterases releasing the fluorescent Fluo-3 dye inside cells Once inside Fluo-3 binds to free calcium ions resulting in a fluorescent signal that correlates with calcium levels This assay is especially useful in in vitro studies often demonstrating activity in the nanomolar to low micromolar range Typical applications include investigating calcium signaling pathways in cell models screening potential drug effects and facilitating animal studies Researchers employ this kit for its ability to provide real-time insights into calcium dynamics in various biological systems Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67948 Supplier Apexbio Technology LLC Supplier No. K2299200
Fluo-3 AM Calcium Assay Kit is a widely used tool in biomedical research for monitoring calcium ion concentrations in various cellular processes Originating from the category of fluorescent calcium indicators this kit operates by permeating cell membranes and then being hydrolyzed by intracellular esterases releasing the fluorescent Fluo-3 dye inside cells Once inside Fluo-3 binds to free calcium ions resulting in a fluorescent signal that correlates with calcium levels This assay is especially useful in in vitro studies often demonstrating activity in the nanomolar to low micromolar range Typical applications include investigating calcium signaling pathways in cell models screening potential drug effects and facilitating animal studies Researchers employ this kit for its ability to provide real-time insights into calcium dynamics in various biological systems Experimental concentrations typically depend on the specific research design