Learn More
Biotium Fluo-3, AM Ester, 10x100ug
Catalog No. 501964685
Encompass_Preferred
Membrane-permeant AM ester form of Fluo-3 that can be loaded into cells via incubation. Because of the relatively low water solubility of the AM ester, Pluronic F-127, a mild detergent, is often used as a dispersing agent to facilitate dye loading. Fluo-3 AM ester itself does not bind Ca2 , but it is readily hydrolyzed to Fluo-3 by endogenous esterases once the dye is inside the cells. An important application of Fluo-3 AM ester is its use in high throughput drug screening. Properties: Orange red solid soluble in DMSO; Store -20°C and protect from light, especially when in solution; C51H50Cl2N2O23; MW: 1129.9; [121714-22-5]
Catalog No. 50-196-4685 Supplier Biotium Supplier No. 50013
Membrane-permeant AM ester form of Fluo-3 that can be loaded into cells via incubation. Because of the relatively low water solubility of the AM ester, Pluronic F-127, a mild detergent, is often used as a dispersing agent to facilitate dye loading. Fluo-3 AM ester itself does not bind Ca2 , but it is readily hydrolyzed to Fluo-3 by endogenous esterases once the dye is inside the cells. An important application of Fluo-3 AM ester is its use in high throughput drug screening. Properties: Orange red solid soluble in DMSO; Store -20°C and protect from light, especially when in solution; C51H50Cl2N2O23; MW: 1129.9; [121714-22-5]