Learn More
Abcam Fluo-3 AM, Fluorescent calcium Indicator, 1MG
Catalog No. 502705357
Encompass_Preferred
MW 1129.85 g/mol, Purity >95%. Cell-permeable acetoxymethyl (AM) ester of the fluorescent calcium indicator Fluo-3. Commonly used in cell-based experiments to detect changes in intracellular calcium levels. Practically nonfluorescent until it is hydrolyzed intracellularly to Fluo-3 and in the presence of Ca²+.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5357 Supplier Abcam Supplier No. AB1452541MG
MW 1129.85 g/mol, Purity >95%. Cell-permeable acetoxymethyl (AM) ester of the fluorescent calcium indicator Fluo-3. Commonly used in cell-based experiments to detect changes in intracellular calcium levels. Practically nonfluorescent until it is hydrolyzed intracellularly to Fluo-3 and in the presence of Ca²+.
The product is subject to the following: Abcam Restricted Use Statement