Learn More
Enzo Life Sciences Fluo-3 AM (ultra pure) (1mg). CAS: 121714-22-5
SDP
Supplier:  Enzo Life Sciences ENZ52004
Encompass_Preferred
Fluo-3 AM is widely used in flow cytometry, confocal laserscanning microscopy and fluorescence spectroscopy. The probe has proved particularly valuable for determining GPCR function via cell-based high-throughput screening. Fluo-3 AM is a membrane permeable probe and can be passively loaded in cells by simple incubation. Once inside the cell, Fluo-3 becomes fluorescent in the presence of free Ca (II) . Fluo-3 is a long wavelength calcium indicator probe (488 nm excitable). The longer wavelength of absorption is convenient for minimizing photodamage in cells. Wavelength Maxima: Excitation 506nm, Emission 526nm Alternative name: 1-[Amino-5-(2,7-dichloro-6-acetomethoxy-3-oxo-3H-xanthen-9-yl)phenoxy]-2-(2'-amino-5'-methylphenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxy-methyl ester. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-2603