Learn More
Medchemexpress LLC Fluo-3AM (Fluo-3-pentaacetoxymethyl ester) | 121714-22-5 | ≥99.0% | 500 UG
SDP
Catalog No. 5000410357
Encompass_Preferred
Fluo-3 AM is a fluorescent Ca2+ chelator with high affinity for calcium. It can specifically identify intracellular calcium ions, offering high sensitivity and low cytotoxicity. The increased AM acetylmethyl ester allows it to enter cells effectively. Once inside, it is sheared by intracellular esterase and remains in the cell to bind with calcium ions, producing strong fluorescence. It is suitable for laboratory centrifugation and storage.
  • High affinity for calcium
  • High sensitivity
  • Low cytotoxicity
  • Effective cell entry due to increased AM acetylmethyl ester
  • Binds with calcium ions, producing strong fluorescence
  • Green fluorescence calcium indicator
  • Useful in calcium flux analysis
Catalog No. 50-004-10357 Supplier Medchemexpress LLC Supplier No. HYD0716500UG
Fluo-3 AM is a fluorescent Ca2+ chelator with high affinity for calcium. It can specifically identify intracellular calcium ions, offering high sensitivity and low cytotoxicity. The increased AM acetylmethyl ester allows it to enter cells effectively. Once inside, it is sheared by intracellular esterase and remains in the cell to bind with calcium ions, producing strong fluorescence. It is suitable for laboratory centrifugation and storage.
  • High affinity for calcium
  • High sensitivity
  • Low cytotoxicity
  • Effective cell entry due to increased AM acetylmethyl ester
  • Binds with calcium ions, producing strong fluorescence
  • Green fluorescence calcium indicator
  • Useful in calcium flux analysis