Learn More
Medchemexpress LLC Fluo-4 AM (acetoxymethyl ester) | 273221-67-3 | MFCD18837018 | >=99.0% | 1096.94 | C51H50F2N2O23 | 500 UG
SDP
Catalog No. 5000246529
Encompass_Preferred
Fluo-4 AM is a cell-permeable fluorescent calcium indicator (acetoxymethyl ester) that is hydrolyzed by intracellular esterases to release Fluo-4, enabling sensitive detection of intracellular Ca2+ changes (Ex/Em = 485/526 nm). Supplied as a solid, it is unstable in solution and requires fresh stock and working solutions prepared and protected from light.
  • High purity: ≥99.0% as stated by manufacturer.
  • Bright green fluorescence with excitation 485 nm and emission 526 nm.
  • Cell permeable and cleaved intracellularly to report intracellular Ca2+.
  • Suitable for live-cell calcium imaging and assays.
  • Supplied as a solid for weigh-and-dissolve preparation of stock solutions.
  • Store at -20°C, sealed, protected from moisture and light.
Catalog No. 50-002-46529 Supplier Medchemexpress LLC Supplier No. HY101896500UG
Add to Cart
Add to Cart
Fluo-4 AM is a cell-permeable fluorescent calcium indicator (acetoxymethyl ester) that is hydrolyzed by intracellular esterases to release Fluo-4, enabling sensitive detection of intracellular Ca2+ changes (Ex/Em = 485/526 nm). Supplied as a solid, it is unstable in solution and requires fresh stock and working solutions prepared and protected from light.
  • High purity: ≥99.0% as stated by manufacturer.
  • Bright green fluorescence with excitation 485 nm and emission 526 nm.
  • Cell permeable and cleaved intracellularly to report intracellular Ca2+.
  • Suitable for live-cell calcium imaging and assays.
  • Supplied as a solid for weigh-and-dissolve preparation of stock solutions.
  • Store at -20°C, sealed, protected from moisture and light.