Learn More
Cayman Chemical Fluo-4 (potassium salt), Purity: ≥90%, CAS: 1199351-32-0, 0.5 mg
SDP
Catalog No. 502911202
Encompass_Preferred
A cell-impermeable fluorescent calcium indicator; binds to calcium (Kd = 355 nM); ex/em = 490/515 nm
Catalog No. 50-291-1202 Supplier Cayman Chemical Supplier No. 357650.5 MG
A cell-impermeable fluorescent calcium indicator; binds to calcium (Kd = 355 nM); ex/em = 490/515 nm