Learn More
Aat Bioquest Fluo-5N AM Cell permeant (10x50 ug)
SDP
Supplier:  Aat Bioquest 20566
Encompass_Preferred
Fluo-5N is an analog of Fluo-4 with lower calcium-binding affinity Kd 90 uM making it suitable for detecting intracellular calcium levels in the range of 1 microM to 1 mM that would saturate the response of Fluo-4. This catalog is for 10x50 ug.
Catalog No. 50-224-9866