Learn More
Selleck Chemical LLC Fluocinolone Acetonide 100mg 67-73-2 Flucort-N
SDP
Catalog No. 501362444
Encompass_Preferred
Fluocinolone Acetonide (Flucort-N) is a corticosteroid that binds to the cytosolic glucocorticoid receptor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2444 Supplier Selleck Chemical LLC Supplier No. S2470100MG
Fluocinolone Acetonide (Flucort-N) is a corticosteroid that binds to the cytosolic glucocorticoid receptor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.