Learn More
Apexbio Technology LLC Fluocinolone Acetonide 67-73-2 200mg
SDP
Catalog No. 5000091013
Encompass_Preferred
Fluocinolone Acetonide (67-73-2) is a glucocorticoid receptor (GR) agonist targeting the GR It is designed to modulate intracellular signaling pathways mediated by the GR thereby regulating inflammatory response and influencing lipid metabolism by reducing lipid accumulation in tissues Fluocinolone Acetonide exerts its biological activity primarily through activation of the GR In cellular assays Fluocinolone Acetonide demonstrates stimulation of proliferation in dental pulp cells (DPCs) Based on these pharmacological properties Fluocinolone Acetonide holds research potential in studies of dental tissue regeneration and repair as well as in investigations of preventative and therapeutic strategies for chemotherapy-induced peripheral neuropathy
Catalog No. 50-000-91013 Supplier Apexbio Technology LLC Supplier No. B2095200
Fluocinolone Acetonide (67-73-2) is a glucocorticoid receptor (GR) agonist targeting the GR It is designed to modulate intracellular signaling pathways mediated by the GR thereby regulating inflammatory response and influencing lipid metabolism by reducing lipid accumulation in tissues Fluocinolone Acetonide exerts its biological activity primarily through activation of the GR In cellular assays Fluocinolone Acetonide demonstrates stimulation of proliferation in dental pulp cells (DPCs) Based on these pharmacological properties Fluocinolone Acetonide holds research potential in studies of dental tissue regeneration and repair as well as in investigations of preventative and therapeutic strategies for chemotherapy-induced peripheral neuropathy