Learn More
Medchemexpress LLC Fluocinolone (Acetonide) | 67-73-2 | 99.68% | 1 ML
SDP
Catalog No. 5000407040
Encompass_Preferred
Fluocinolone (Acetonide) is a glucocorticoid receptor agonist. It is effective in preventing both lipid accumulation and inflammation. This solution can promote the proliferation of DPCs and has the potential role in repairing injured pulp tissues. It can also be used to study the prevention of chemotherapy-induced peripheral neuropathy.
  • Effective in preventing lipid accumulation and inflammation
  • Promotes proliferation of DPCs
  • Potential role in repairing injured pulp tissues
  • Can be used to study prevention of chemotherapy-induced peripheral neuropathy
  • Improves foam cell survival in THP-1 cells
  • Inhibits inflammatory cytokine secretion and reduces cholesteryl ester accumulation
Catalog No. 50-004-07040 Supplier Medchemexpress LLC Supplier No. HYB041510MM/1ML
Fluocinolone (Acetonide) is a glucocorticoid receptor agonist. It is effective in preventing both lipid accumulation and inflammation. This solution can promote the proliferation of DPCs and has the potential role in repairing injured pulp tissues. It can also be used to study the prevention of chemotherapy-induced peripheral neuropathy.
  • Effective in preventing lipid accumulation and inflammation
  • Promotes proliferation of DPCs
  • Potential role in repairing injured pulp tissues
  • Can be used to study prevention of chemotherapy-induced peripheral neuropathy
  • Improves foam cell survival in THP-1 cells
  • Inhibits inflammatory cytokine secretion and reduces cholesteryl ester accumulation