Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluocinolone acetonide, ≥97.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015143 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5143 100 mg
11-101-5142 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5143 Supplier MilliporeSigma Supelco Supplier No. F8880100MG
Add to Cart
Add to Cart

Specifications

CAS 67-73-2
Physical Form Neat
Linear Formula C24H30F2O6
MDL Number MFCD00010525
Melting Point 267°C to 269°C
Synonym 6 alpha,9 alpha-Difluoro-11 beta,16 alpha,17 alpha,21-tetrahydroxy-1,4-pregnadiene-3,20-dione; 6 alpha,9 alpha-Difluoro-16 alpha-hydroxyprednisolone 16,17-acetonide; 6 alpha-Fluorotriamcinolone acetonide; Sinalar; Synandone
Type Analytical Standard
Quantity 100 mg
Formula Weight 452.49
Percent Purity ≥97.5% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.