Learn More
Enzo Life Sciences GFP-CERTIFIED FLUOFORTE Calcium assay kit (100x96wells)
SDP
Supplier:  Enzo Life Sciences ENZ51020KP100
Encompass_Preferred
Enzo Life Sciences’ GFP-CERTIFIED FLUOFORTE Calcium Assay Kit (high-throughput) provides a homogeneous fluorescence-based assay for detecting intracellular calcium mobilization across a broad spectrum of biological targets. Relative to other commercially available dyes, the GFP-CERTIFIED FLUOFORTE dye is the brightest and most sensitive fluorescent calcium indicator. The kit provides a homogeneous mix-and-read, no-wash calcium mobilization assay. The homogenous cell-based assay for calcium offers fewer steps, lower variability and an easier protocol for adherent and non-adherent cell lines. In addition, it requires neither a washing step, nor exogenous addition of a quencher dye, which could adversely effect receptor-ligand interaction kinetics. Applications: Flow Cytometry, Fluorescence microscopy, Fluorescent detection, HTS .
Catalog No. 50-201-2592