Learn More
Enzo Life Sciences FLUOFORTE Calcium assay kit (10x96tests)
SDP
Catalog No. NC9001851
Encompass_Preferred
A Brighter, More Robust Fluorescent Assay for Calcium Mobilization. Applications: Flow Cytometry. Fluorescence microscopy. Fluorescent detection, HTS. Contents: Reagent A (lyophilized FLUOFORTE dye), 1 vial. Reagent B (dye efflux inhibitor), 10 x 1 ml. Reagent C (Hanks’ buffer with 20mm HEPES), 100 ml
Catalog No. NC9001851 Supplier Enzo Life Sciences Supplier No. ENZ51017
A Brighter, More Robust Fluorescent Assay for Calcium Mobilization. Applications: Flow Cytometry. Fluorescence microscopy. Fluorescent detection, HTS. Contents: Reagent A (lyophilized FLUOFORTE dye), 1 vial. Reagent B (dye efflux inhibitor), 10 x 1 ml. Reagent C (Hanks’ buffer with 20mm HEPES), 100 ml