Learn More
Enzo Life Sciences FLUOFORTE Reagent (1mg)
SDP
Supplier:  Enzo Life Sciences ENZ52015
Encompass_Preferred
Used in a homogeneous fluorescence-based assay for detecting intracellular calcium mobilization across a broad spectrum of biological targets. Fluorescent calcium indicator dye developed to improve cell loading and calcium response while maintaining the convenient Fluo-3, Fluo-4 and Calcium 4 spectral wavelength of maximum excitation at ~490 nm and maximum emission at ~520 nm. Relative to the cited dyes, FLUOFORTE AM dye yields the brightest signal and largest assay window. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C. Applications: Flow Cytometry, Fluorescence microscopy, Fluorescent detection, HTS .
Catalog No. 50-201-2610