Learn More
Medchemexpress LLC Fluopimomide | 1309859-39-9 | ≥97.0% | 416.68 | C15H8ClF7N2O2 | 250 MG
SDP
Catalog No. 5000281179
Encompass_Preferred
Fluopimomide is a potent pesticide used in agricultural pest management and research applications. It adversely affects nematode growth, locomotion, reproduction, and lifespan, in part by inducing reactive oxygen species and inhibiting antioxidant systems. Supplied as a solid with defined purity and storage recommendations, it is intended for use in formulation development and biological studies.
  • Potent activity against nematodes and certain plant pathogens.
  • Disrupts growth, locomotion, reproduction, and lifespan via ROS generation.
  • Inhibits nematode antioxidant systems.
  • High purity (≥97.0%) suitable for research and analytical use.
  • Solid, white to off-white appearance for handling and formulation.
  • Recommended storage under nitrogen; solvent stability data provided.
Catalog No. 50-002-81179 Supplier Medchemexpress LLC Supplier No. HYW420337250MG
Fluopimomide is a potent pesticide used in agricultural pest management and research applications. It adversely affects nematode growth, locomotion, reproduction, and lifespan, in part by inducing reactive oxygen species and inhibiting antioxidant systems. Supplied as a solid with defined purity and storage recommendations, it is intended for use in formulation development and biological studies.
  • Potent activity against nematodes and certain plant pathogens.
  • Disrupts growth, locomotion, reproduction, and lifespan via ROS generation.
  • Inhibits nematode antioxidant systems.
  • High purity (≥97.0%) suitable for research and analytical use.
  • Solid, white to off-white appearance for handling and formulation.
  • Recommended storage under nitrogen; solvent stability data provided.