Learn More
Medchemexpress LLC HY-119459 50mg Medchemexpress, Fluopyram CAS:658066-35-4 Purity:>98%
SDP
Catalog No. 501969538
Encompass_Preferred
Medchemexpress, HY-119459 50mg Fluopyram CAS:658066-35-4 Fluopyram is a succinate dehydrogenase inhibitor fungicide, inhibits the growth of F. virguliforme isolates with mean EC50 of 3.35 µg/mL. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9538 Supplier Medchemexpress LLC Supplier No. HY11945950MG
Medchemexpress, HY-119459 50mg Fluopyram CAS:658066-35-4 Fluopyram is a succinate dehydrogenase inhibitor fungicide, inhibits the growth of F. virguliforme isolates with mean EC50 of 3.35 µg/mL. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.