Learn More
Molecular Devices Molecular Devices Reporter Assay Kit, Dual Luciferase, Flash, 50 Reactions
Catalog No. 5000014560
Encompass_Preferred
The SpectraMax DuoLuc Reporter Assay Kit is an evaluation kit for sensitive, sequential measurement of firefly and Renilla luciferase activity in mammalian cells. The kit uses flash-type luminescence and is optimized for microplate readers with injector capability, providing reagents for cell lysis and substrate reactions for approximately 50 assays.
  • Enables sequential measurement of firefly and Renilla luciferase using flash-type luminescence.
  • Provides reagents for approximately 50 reactions.
  • Optimized for use on microplate readers with injector capability.
  • Typical workflow uses about 20 μL of cell lysate per well and 100 μL injection volumes for each substrate.
  • Includes reagents for cell lysis and substrate preparation to support rapid assay setup.
Catalog No. 50-000-14560 Supplier Molecular Devices Supplier No. R8360
The SpectraMax DuoLuc Reporter Assay Kit is an evaluation kit for sensitive, sequential measurement of firefly and Renilla luciferase activity in mammalian cells. The kit uses flash-type luminescence and is optimized for microplate readers with injector capability, providing reagents for cell lysis and substrate reactions for approximately 50 assays.
  • Enables sequential measurement of firefly and Renilla luciferase using flash-type luminescence.
  • Provides reagents for approximately 50 reactions.
  • Optimized for use on microplate readers with injector capability.
  • Typical workflow uses about 20 μL of cell lysate per well and 100 μL injection volumes for each substrate.
  • Includes reagents for cell lysis and substrate preparation to support rapid assay setup.