Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Devices Molecular Devices dsDNA Assay Kit, Fluorescence, 350/460 nm, 2-2,000 ng

Catalog No. 5000014455
Encompass_Preferred

SpectraMax Quant AccuBlue HiRange dsDNA Assay Kit provides fluorescence-based quantitation of double-stranded DNA across a broad dynamic range. The kit is supplied with reagents sufficient for two 96-well plates and is configured for microplate-reader workflows.

  • Fluorescence-based assay for dsDNA quantitation.
  • Detection range 2 ng-2,000 ng per assay.
  • Excitation 350 nm, emission 460 nm.
  • Sufficient reagents for two 96-well plates.
  • High selectivity for double-stranded DNA.

Catalog No. 50-000-14455 Supplier Molecular Devices Supplier No. R8358
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SpectraMax Quant AccuBlue HiRange dsDNA Assay Kit provides fluorescence-based quantitation of double-stranded DNA across a broad dynamic range. The kit is supplied with reagents sufficient for two 96-well plates and is configured for microplate-reader workflows.

  • Fluorescence-based assay for dsDNA quantitation.
  • Detection range 2 ng-2,000 ng per assay.
  • Excitation 350 nm, emission 460 nm.
  • Sufficient reagents for two 96-well plates.
  • High selectivity for double-stranded DNA.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.