Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Devices Molecular Devices dsDNA Assay Kit, Fluorescence, 30-250 ng

Catalog No. 5000014369
Encompass_Preferred

The SpectraMax Quant AccuClear Nano dsDNA Assay Kit is a fluorescence-based assay for selective quantitation of double-stranded DNA in microplate format. The Explorer kit includes reagents and standards sufficient for two 96-well plates and provides a detection range of 30 pg to 250 ng per well with preconfigured protocols for plate reader software.

  • Fluorescence-based dsDNA quantitation.
  • Sensitivity range 30 pg to 250 ng per well.
  • Reagents sufficient for two 96-well plates.
  • Includes dye, enhancer, buffer, and DNA standards.
  • High selectivity for double-stranded DNA over single-stranded DNA and RNA.
  • Excitation 468 nm, emission 507 nm optical settings.
  • Optimized for microplate readers with preconfigured protocols in plate reader software.

Catalog No. 50-000-14369 Supplier Molecular Devices Supplier No. R8356
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The SpectraMax Quant AccuClear Nano dsDNA Assay Kit is a fluorescence-based assay for selective quantitation of double-stranded DNA in microplate format. The Explorer kit includes reagents and standards sufficient for two 96-well plates and provides a detection range of 30 pg to 250 ng per well with preconfigured protocols for plate reader software.

  • Fluorescence-based dsDNA quantitation.
  • Sensitivity range 30 pg to 250 ng per well.
  • Reagents sufficient for two 96-well plates.
  • Includes dye, enhancer, buffer, and DNA standards.
  • High selectivity for double-stranded DNA over single-stranded DNA and RNA.
  • Excitation 468 nm, emission 507 nm optical settings.
  • Optimized for microplate readers with preconfigured protocols in plate reader software.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.