Learn More
Azzota Corp Fluor Cuvette 10mm 1.4ml Qz
Catalog No. 5000887136
Encompass_Preferred
Quartz Fluorometer Cuvettes Four (4) sides clear 10mm Pathlength 4mm Inside width 1 40ml Volume Dimension 45x12 5x12 5mm
Catalog No. 50-008-87136 Supplier Azzota Corp Supplier No. Q534
Quartz Fluorometer Cuvettes Four (4) sides clear 10mm Pathlength 4mm Inside width 1 40ml Volume Dimension 45x12 5x12 5mm