Learn More
Azzota Corp Fluor cuvette 10mm 1.7ml round
Catalog No. 5000887011
Encompass_Preferred
FLUOROMETER CUVETTE 10MM PATHLENGTH 1 7ML Four (4) sides clear Pathlength - 10mm Inside width - 5mm Volume - 1 7ml Dimension - 45x12 5x12 5mm Round Bottom Includes Cover Type G - Optical Glass (340nm-2500nm)
Catalog No. 50-008-87011 Supplier Azzota Corp Supplier No. G544
FLUOROMETER CUVETTE 10MM PATHLENGTH 1 7ML Four (4) sides clear Pathlength - 10mm Inside width - 5mm Volume - 1 7ml Dimension - 45x12 5x12 5mm Round Bottom Includes Cover Type G - Optical Glass (340nm-2500nm)