Learn More
Azzota Corp Fluor cuvette 4win 10mm 0.7ml
Catalog No. 5000887009
Encompass_Preferred
Glass Fluorometer Cuvettes (340nm-2500nm) Four (4) sides clear 10mm Pathlength 2mm Inside width 0 70ml Volume Dimension 45x12 5x12 5mm
Catalog No. 50-008-87009 Supplier Azzota Corp Supplier No. G514
Glass Fluorometer Cuvettes (340nm-2500nm) Four (4) sides clear 10mm Pathlength 2mm Inside width 0 70ml Volume Dimension 45x12 5x12 5mm