Learn More
Azzota Corp Fluor cuvette 4win 10mm 1.0ml
Catalog No. 5000887010
Encompass_Preferred
Glass Fluorometer Cuvettes (340nm-2500nm) Four (4) sides clear 10mm Pathlength 3mm Inside width 1 00ml Volume Dimension 45x12 5x12 5mm
Catalog No. 50-008-87010 Supplier Azzota Corp Supplier No. G524
Glass Fluorometer Cuvettes (340nm-2500nm) Four (4) sides clear 10mm Pathlength 3mm Inside width 1 00ml Volume Dimension 45x12 5x12 5mm