Learn More
Enzo Life Sciences FLUOR DE LYS HDAC fluorometric cellular activity assay kit (1x96wells)
SDP
Catalog No. NC1372671
Encompass_Preferred
Monitor HDAC activity in cell culture using cell-permeable FLUOR DE LYS substrate. Alternative Name: Cellular Histone deacetylase fluorescent assay kit. Applications: Fluorescent detection, HTS, Cell-based assays, Activity assay. Contents: Nuclear Extracts from Hela Cells (human cervical cancer cell line) (BML-KI345), FLUOR DE LYS Substrate (BML-KI104), FLUOR DE LYS Developer Concentrate (20x), Trichostatin A (HDAC Inhibitor) (BML-GR-309-9090), Nicotinamide (Sirtuin Inhibitor) (BML-KI283), FLUOR DE LYS Deacetylated Standard (BML-KI142), HDAC Assay Buffer (BML-KI143), Cell Lysis Buffer (2), 1/2 volume microplates (BML-KI101).
Catalog No. NC1372671 Supplier Enzo Life Sciences Supplier No. BMLAK5030001
Monitor HDAC activity in cell culture using cell-permeable FLUOR DE LYS substrate. Alternative Name: Cellular Histone deacetylase fluorescent assay kit. Applications: Fluorescent detection, HTS, Cell-based assays, Activity assay. Contents: Nuclear Extracts from Hela Cells (human cervical cancer cell line) (BML-KI345), FLUOR DE LYS Substrate (BML-KI104), FLUOR DE LYS Developer Concentrate (20x), Trichostatin A (HDAC Inhibitor) (BML-GR-309-9090), Nicotinamide (Sirtuin Inhibitor) (BML-KI283), FLUOR DE LYS Deacetylated Standard (BML-KI142), HDAC Assay Buffer (BML-KI143), Cell Lysis Buffer (2), 1/2 volume microplates (BML-KI101).