Learn More
Enzo Life Sciences FLUOR DE LYS HDAC6 fluorometric drug discovery kit (1x96wells)
SDP
Catalog No. NC1646443
Encompass_Preferred
A FLUOR DE LYS. HDAC6 assay is based on the FLUOR DE LYS Substrate and FLUOR DE LYS Developer combination. The assay procedure has two steps. First, the FLUOR DE LYS Substrate, which comprises an acetylated lysine side chain, is incubated with HDAC6. Deacetylation of the substrate sensitizes the substrate so that, in the second step, treatment with the FLUOR DE LYSDeveloper produces a fluorophore. Alternative Name: HDAC6 fluorescent assay kit. Applications: Cell-based assays, Activity assay.
Catalog No. NC1646443 Supplier Enzo Life Sciences Supplier No. BMLAK5160001
A FLUOR DE LYS. HDAC6 assay is based on the FLUOR DE LYS Substrate and FLUOR DE LYS Developer combination. The assay procedure has two steps. First, the FLUOR DE LYS Substrate, which comprises an acetylated lysine side chain, is incubated with HDAC6. Deacetylation of the substrate sensitizes the substrate so that, in the second step, treatment with the FLUOR DE LYSDeveloper produces a fluorophore. Alternative Name: HDAC6 fluorescent assay kit. Applications: Cell-based assays, Activity assay.