Learn More
Enzo Life Sciences FLUOR DE LYS SIRT3 fluorometric drug discovery assay kit (1x96wells)
SDP
Catalog No. NC1370353
Encompass_Preferred
Alternative Name: Sirtuin 3 fluorescent assay kit. Applications: Fluorescent detection, HTS, Cell-based assays, Activity assay. Contents: SIRT3 (Sirtuin 3) (human, recombinant) (BML-SE270), FLUOR DE LYS-SIRT2 substrate (BML-KI179) 500µM NAD; Recombinant enzyme dissolved in See vial label for activity and protein. Concentrations FLUOR DE LYS-SIRT2, Deacetylase substrate (BML-KI179) Note: this SIRT2 substrate is also hydolyzed by SIRT3. FLUOR DE LYS Developer II Concentrate (5x) (BML-KI176) NAD (Sirtuin Substrate) (BML-KI282) Nicotinamide (Sirtuin Inhibitor) (BML-KI283) (500µl; 5 mM Nicotinamide in 50mM TRIS/Cl, pH 8.0, 137mM sodium chloride, 2.7mM potassium chloride, 1mM magnesium chloride) Suramin sodium (Sirtuin Inhibitor) (BML-KI285) (10 mg; Solid. MW: 1429.2, soluble in water or assay buffer (to 25mM)) FLUOR DE LYS Deacetylated Standard (BML-KI142) Sirtuin Assay Buffer (BML-KI286) 1/2 volume microplate (BML-KI101) 1/2 volume white microplate (BML-KI110)
Catalog No. NC1370353 Supplier Enzo Life Sciences Supplier No. BMLAK5570001
Alternative Name: Sirtuin 3 fluorescent assay kit. Applications: Fluorescent detection, HTS, Cell-based assays, Activity assay. Contents: SIRT3 (Sirtuin 3) (human, recombinant) (BML-SE270), FLUOR DE LYS-SIRT2 substrate (BML-KI179) 500µM NAD; Recombinant enzyme dissolved in See vial label for activity and protein. Concentrations FLUOR DE LYS-SIRT2, Deacetylase substrate (BML-KI179) Note: this SIRT2 substrate is also hydolyzed by SIRT3. FLUOR DE LYS Developer II Concentrate (5x) (BML-KI176) NAD (Sirtuin Substrate) (BML-KI282) Nicotinamide (Sirtuin Inhibitor) (BML-KI283) (500µl; 5 mM Nicotinamide in 50mM TRIS/Cl, pH 8.0, 137mM sodium chloride, 2.7mM potassium chloride, 1mM magnesium chloride) Suramin sodium (Sirtuin Inhibitor) (BML-KI285) (10 mg; Solid. MW: 1429.2, soluble in water or assay buffer (to 25mM)) FLUOR DE LYS Deacetylated Standard (BML-KI142) Sirtuin Assay Buffer (BML-KI286) 1/2 volume microplate (BML-KI101) 1/2 volume white microplate (BML-KI110)