Learn More
Innovative Research Inc Fluor lbl Phe-Pro-Arg-Chloro
Catalog No. 502744612
Encompass_Preferred
Fluorescein Labeled Phe-Pro-Arg-Chloromethylketone 1MG - Fluorescein Labeled Phe-Pro-Arg-Chloromethylketone from Innovative Research is a member of the tri-peptide chloromethylketone family of reagents, which have seen extensive application in the irreversible inhibition of various serine proteases. Innovative Research offers both labeled and unlabeled species of GGACK and PPACK, also known as EGRCK and FPRCK, respectively. This product is offered in a frozen liquid format, with a purity of 95% by HPLC analysis.
Catalog No. 50-274-4612 Supplier Innovative Research Inc Supplier No. ICMPPACKFITC1MG
Fluorescein Labeled Phe-Pro-Arg-Chloromethylketone 1MG - Fluorescein Labeled Phe-Pro-Arg-Chloromethylketone from Innovative Research is a member of the tri-peptide chloromethylketone family of reagents, which have seen extensive application in the irreversible inhibition of various serine proteases. Innovative Research offers both labeled and unlabeled species of GGACK and PPACK, also known as EGRCK and FPRCK, respectively. This product is offered in a frozen liquid format, with a purity of 95% by HPLC analysis.