Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoranthene, MilliporeSigma™ Supelco™

Catalog No. 111020078 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0078 5000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0078 Supplier MilliporeSigma Supelco Supplier No. 48535
Add to Cart
Add to Cart

FA, a six ringed highly stable compound, is considered as an environmental indicator. It is mainly selected for simulation study.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 206-44-0
Flash Point 198°C
Physical Form Neat
Linear Formula C16H10
MDL Number MFCD00001184
Melting Point 105°C to 110°C
Boiling Point 384°C
Synonym Benzo[j,k]fluorene
Type Organic Pollutant Standard
Quantity 5000 mg
Formula Weight 202.25
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.