Learn More
Medchemexpress LLC HY-W026772 500mg Medchemexpress, Fluorene CAS:86-73-7 Purity:>98%
SDP
Catalog No. 502083444
Encompass_Preferred
Medchemexpress, HY-W026772 500mg Fluorene CAS:86-73-7 Fluorene, a polycyclic aromatic hydrocarbon (PAH), is a precursor to other fluorene compounds. Fluorene and its derivative can be used as a precursor to pharmaceuticals or fluorene-based dyes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3444 Supplier Medchemexpress LLC Supplier No. HYW026772500MG
Medchemexpress, HY-W026772 500mg Fluorene CAS:86-73-7 Fluorene, a polycyclic aromatic hydrocarbon (PAH), is a precursor to other fluorene compounds. Fluorene and its derivative can be used as a precursor to pharmaceuticals or fluorene-based dyes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.