Learn More
Medchemexpress LLC Fluorene | 86-73-7 | MFCD00001111 | 98.1% | 166.22 g/mol | C13H10 | 1 ML
SDP
Catalog No. 5000301283
Encompass_Preferred
Fluorene (CAS 86-73-7) is a polycyclic aromatic hydrocarbon commonly used as a research chemical and analytical standard. It is supplied as solid material in multiple masses and as a pre-made 10 mM solution in DMSO for laboratory use. Handle according to safety data sheets and standard laboratory precautions, protecting from light where indicated.
  • Polycyclic aromatic hydrocarbon for research and analytical use.
  • Available as solid (multiple sizes) and 10 mM solution in DMSO.
  • High purity suitable for analytical applications.
  • DMSO-soluble with recommended ultrasonic assistance if needed.
  • Store in solvent at low temperature; protect solid from light.
Catalog No. 50-003-01283 Supplier Medchemexpress LLC Supplier No. HYW02677210MM/1ML
Fluorene (CAS 86-73-7) is a polycyclic aromatic hydrocarbon commonly used as a research chemical and analytical standard. It is supplied as solid material in multiple masses and as a pre-made 10 mM solution in DMSO for laboratory use. Handle according to safety data sheets and standard laboratory precautions, protecting from light where indicated.
  • Polycyclic aromatic hydrocarbon for research and analytical use.
  • Available as solid (multiple sizes) and 10 mM solution in DMSO.
  • High purity suitable for analytical applications.
  • DMSO-soluble with recommended ultrasonic assistance if needed.
  • Store in solvent at low temperature; protect solid from light.