Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorene-9-carboxylic acid, 97%

Catalog No. AAB2322422
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 1989-33-9 | C14H10O2 | 210.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1989-33-9
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.232
MDL Number MFCD00001136
InChI Key DNVJGJUGFFYUPT-UHFFFAOYSA-N
Synonym fluorene-9-carboxylic acid, 9-fluorenecarboxylic acid, 9-carboxyfluorene, diphenyleneacetic acid, 9-fluorencarboxylic acid, rarechem aq bd 0ba1, akos auf02021, 9-carboxy-9h-fluorene, enamine_003081, 9-fluorenylcarboxylic acid
PubChem CID 74809
IUPAC Name 9H-fluorene-9-carboxylic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)C(=O)O

Specifications

Melting Point 226°C to 232°C
Flash Point 230°C (446°F)
Quantity 100 g
Beilstein 1911728
Formula Weight 210.23
Percent Purity 97%
Chemical Name or Material Fluorene-9-carboxylic acid
EINECSNumber 217-866-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.