Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fluorene-d10 | 81103-79-9 | MFCD00144313 | 98.0% | 176.29 g/mol | C13D10 | 50 MG
SDP

Catalog No. 5000301749
Encompass_Preferred

Fluorene-d10 is the deuterium-labeled isotopologue of fluorene supplied as a research-grade analytical standard. It is used in mass spectrometry, tracer studies, and as a labeled precursor in synthetic chemistry. The material is a white to off-white solid with high isotopic enrichment and is intended for research use only.

  • Deuterium-labeled fluorene for tracer and analytical work.
  • High purity: 98.0% (HPLC).
  • Molecular formula C13D10; molecular weight 176.29 g/mol.
  • White to off-white solid; soluble in DMSO and ethanol.
  • Store at -20°C and protect from light.
  • Intended for research use only.

Catalog No. 50-003-01749 Supplier Medchemexpress LLC Supplier No. HYW026772S50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorene-d10 is the deuterium-labeled isotopologue of fluorene supplied as a research-grade analytical standard. It is used in mass spectrometry, tracer studies, and as a labeled precursor in synthetic chemistry. The material is a white to off-white solid with high isotopic enrichment and is intended for research use only.

  • Deuterium-labeled fluorene for tracer and analytical work.
  • High purity: 98.0% (HPLC).
  • Molecular formula C13D10; molecular weight 176.29 g/mol.
  • White to off-white solid; soluble in DMSO and ethanol.
  • Store at -20°C and protect from light.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.