Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fluorene-d10 | 81103-79-9 | MFCD00144313 | 99.0% | 176.28 g/mol | C13D10 | 100 MG
SDP

Catalog No. 5000299704
Encompass_Preferred

Fluorene-d10 is the deuterium-labeled analogue of fluorene (C13D10) used as an analytical standard and internal standard in mass spectrometry and isotope-labeling studies. It is provided as a high-purity material for research and analytical applications, with manufacturer guidance for solubility and formulation.

  • Deuterium-labeled analogue containing 10 deuterium atoms.
  • High purity: 99.0%.
  • Molecular formula C13D10 and molecular weight 176.28 g/mol.
  • Suitable as an internal standard for mass spectrometry and isotope studies.
  • Soluble in DMSO at 50 mg/mL; in vivo formulation guidance available.
  • Offered in small milligram package sizes for analytical use.

Catalog No. 50-002-99704 Supplier Medchemexpress LLC Supplier No. HYW026772S100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorene-d10 is the deuterium-labeled analogue of fluorene (C13D10) used as an analytical standard and internal standard in mass spectrometry and isotope-labeling studies. It is provided as a high-purity material for research and analytical applications, with manufacturer guidance for solubility and formulation.

  • Deuterium-labeled analogue containing 10 deuterium atoms.
  • High purity: 99.0%.
  • Molecular formula C13D10 and molecular weight 176.28 g/mol.
  • Suitable as an internal standard for mass spectrometry and isotope studies.
  • Soluble in DMSO at 50 mg/mL; in vivo formulation guidance available.
  • Offered in small milligram package sizes for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.